Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Procathepsin K, Human, Recombinant Protein, E. coli

Catalog No. 34200110UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-200-110UG 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 34-200-110UG Supplier MilliporeSigma™ Supplier No. 34200110UG
Add to Cart
Add to Cart
Recombinant, human procathepsin K (amino acids 19-329) (GenBank target symbol = S79895, ACC No. P43235) expressed in E. coli with a methionine residue inserted at amino acid 18 to create a new N-terminal initiation site. Cathepsin K, a member of the papain superfamily of cysteine proteinases, plays an important role in osteoclast-mediated bone resorption and collagen degradation. Cathepsin K is synthesized as an inactive proenzyme that is converted to its mature, active form by proteolytic cleavage of the 99 amino acid propeptide domain. Inhibitors of cathepsin K include leupeptin (IC50 = 70nM), E-64 (IC50 = 5nM), and cystatin. Requires activation prior to use.
TRUSTED_SUSTAINABILITY

Specifications

Concentration Concentration on Label
Formulation In 500mM NaCl, 25mM Tris, pH 8.0.
Purification Method SDS-PAGE
Quantity 10 μg
Source E. coli
Immunogen Procathepsin K
Storage Requirements Following initial thaw, aliquot and freeze (−70°C). Following activation the enzyme is unstable and should include MMTS for storage (see recommended reaction conditions for activation).
Regulatory Status RUO
Species Human
Recombinant Recombinant
Form Liquid
Purity or Quality Grade ≥95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.