Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Progard Kit
GSA_VA

Catalog No. PR0GMPK02 Shop All MilliporeSigma Products
Change view
Click to view available options
Polypro HSG 12 in. Cartridge
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
PR0GMPK02 Polypro HSG 12 in. Cartridge
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PR0GMPK02 Supplier MilliporeSigma™ Supplier No. PR0GMPK02
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

With ion-exchange technology

Progard™ 2 Pack & vent filter for Type 2

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Purification Method CaO, Activated Carbon, Anti-scaling, Depth Filtration, Membrane Filter
For Use With (Application) Filters large particles from tap water and reduces hardness prior to reverse osmosis purification
For Use With (Equipment) For Type 2
Includes 1x Progard 2, one 1x Vent Filter for Type 2
Quantity 2 Pack
Type Vent Filter
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.