Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Protein Phosphatase, Lambda, Recombinant Protein, E. coli

Catalog No. 53951420KU Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
20 kU
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-951-420KU 20 kU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 53-951-420KU Supplier MilliporeSigma™ Supplier No. 53951420KU
Add to Cart
Add to Cart
Recombinant λ PPase expressed in E. coli. A 221-amino acid, Mn2+-dependent, protein phosphatase. Hydrolyzes phosphate groups on serine, threonine, tyrosine, or histidine residues. Inhibited by orthovanadate. Note: 1KU = 1,000 units.
TRUSTED_SUSTAINABILITY

Specifications

Formulation In 100mM NaCl, 50mM HEPES, 2mM DTT, 0.1mM MnCl2, 0.1mM EGTA, 50% glycerol, 0.01% BRIJ™ 35 Detergent, pH 7.5.
Preparation Method Further dilute in 50mM Tris-HCl, pH 7.8, 5mM DTT, 2mM MnCl2, and 100μg/mL BSA.
Purification Method SDS-PAGE
Quantity 20 kU
Source E. coli
Immunogen Protein Phosphatase, Lambda
Storage Requirements Following initial thaw, aliquot and freeze (−70°C).
Regulatory Status RUO
Recombinant Recombinant
Form Liquid
Purity or Quality Grade ≥95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.