Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Prothrombin, Human Plasma, Protein

Catalog No. 5395151MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
2 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-951-51MG 1 mg
53-951-52MG 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 53-951-51MG Supplier MilliporeSigma™ Supplier No. 5395151MG
Only null left
Add to Cart
Add to Cart
Native prothrombin from human plasma. Vitamin K-dependent glycoprotein synthesized in the liver. Conversion to thrombin is a key step in the blood coagulation pathway and catalyzes the coagulation of fibrinogen. Clinically, cases of selective deficiency are rare, although, in cases of liver cirrhosis, prothrombin is decreased.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from 100mM NaCl, 20mM Tris-HCl, pH 7.4.
Purification Method SDS-PAGE
Quantity 1 mg
Immunogen Prothrombin
Storage Requirements Following reconstitution, aliquot and freeze (−70°C). Stock solutions are stable for up to 3 months at −70°C.
Regulatory Status RUO
Species Human
Recombinant Native
Form Lyophilized
Purity or Quality Grade ≥95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.