Learn More
MilliporeSigma™ pT7Blue DNA (uncut)
Catalog No. 699673 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
20 μg
Catalog No. Quantity
69-967-3 20 μg
1 options
Catalog No. 69-967-3 Supplier MilliporeSigma™ Supplier No. 699673
Add to Cart
Add to Cart
pT7Blue contains the pUC19 backbone (including high-copy number origin of replication and lac sequences), a T7 promoter, f1 origin of replication, and modified multiple cloning region. The multiple cloning region contains an EcoR V site used for T-cloning flanked by an Nde I site, which allows inserts to be conveniently subcloned into the Nde I site of many pET vectors.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at -20°C. Avoid freeze/thaw cycles.
Quantity 20 μg