Learn More
MilliporeSigma™ pT7Blue T-Vector
Catalog No. 698203 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
69-820-3 1 Kit
1 options
Catalog No. 69-820-3 Supplier MilliporeSigma™ Supplier No. 698203
Add to Cart
Add to Cart
Many thermostable DNA polymerases, such as Taq DNA polymerase, leave single 3' A-nucleotide overhangs on their reaction products. These molecules can be ligated directly to a vector containing compatible single T-nucleotide overhangs. The pT7Blue vector has been specifically constructed for this application and has been prepared for T-cloning by digestion with EcoRV followed by the addition of single 3' dT residues at each end.
TRUSTED_SUSTAINABILITY
Specifications
Promoter T7
Content And Storage Store at -20°C.
Quantity 1 Kit