Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ PTP1B, Human, Recombinant Protein, E. coli

Catalog No. 53973550UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-973-550UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 53-973-550UG Supplier MilliporeSigma™ Supplier No. 53973550UG
Only null left
Add to Cart
Add to Cart
Recombinant, human PTP1B expressed in E. coli. PTP1B, an ubiquitous nontransmembrane protein tyrosine phosphatase, was originally identified in human placenta. This highly active enzyme is useful for the study of tyrosine phosphatase kinetics, substrate specificity, and for screening inhibitors.
TRUSTED_SUSTAINABILITY

Specifications

Concentration Concentration on Label
Formulation In 50mM HEPES, 1mM DTT, 1mM EDTA, 0.05% NP-40, pH 7.2.
Purification Method SDS-PAGE
Quantity 50 μg
Source E. coli
Immunogen PTP1B
Storage Requirements Following initial thaw, aliquot and freeze (−70°C).
Regulatory Status RUO
Species Human
Recombinant Recombinant
Form Liquid
Purity or Quality Grade ≥90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.