Learn More
MilliporeSigma™ pTriEx-4 Ek/LIC Vector Kit
Catalog No. 709053 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
20 Reactions
Catalog No. Quantity
70-905-3 20 Reactions
1 options
Catalog No. 70-905-3 Supplier MilliporeSigma™ Supplier No. 709053
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
The pTriEx™-4 Ek/LIC vector is uniquely designed to allow rapid characterization of target genes in multiple expression systems. With this vector, a single recombinant plasmid can be used to test expression in E. coli, insect and mammalian cells. The pTriEx-4 Ek/LIC vector contains N-terminal His·Tag™ and S·Tag™ coding sequences followed by an enterokinase protease cleavage site and a multiple cloning site, with the option to include a C-terminal His·Tag coding sequence.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at or below -70°C.
Quantity 20 Reactions