Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ PureProteome™ Albumin/IgG Depletion Kit
GSA_VA

Catalog No. LSKMAGD12 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LSKMAGD12 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LSKMAGD12 Supplier MilliporeSigma™ Supplier No. LSKMAGD12
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • PureProteome Albumin/IgG Magnetic Beads, 12mL
  • 10x Phosphate Buffered Saline (PBS) wash and bind buffer, 7mL
  • Amicon Ultra-4 3K Centrifugal Filter, 8/Pk.

Provides rapid, scalable, and reproducible means to deplete >98% of both albumin and IgG from serum and plasma samples, facilitating the detection and analysis of proteins of interest

  • Beads are conjugated to an antibody specific for human serum albumin and to protein G for capturing IgG
  • Adjust bead volume based on sample volume and total protein concentration
  • No manipulation of tubes or multipart assemblies
  • No need for centrifugation; magnetic field immobilizes beads in seconds
  • Particle Size: 10μm
  • Volume: 1 Kit (12mL beads)

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Detectable Analytes Albumin
Type PureProteome™ Albumin/IgG Depletion Kit
For Use With (Application) Serum depletion
Includes Albumin/IgG magnetic beads, 10X Phosphate Buffered Saline (PBS) wash and bind Buffer, Amicon Ultra-4 3K centrifugal filter
Quantity Each
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.