Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ PureProteome™ Carboxy FlexiBind Magnetic Bead System

Catalog No. LSKMAG25C02 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mL
2 x 1mL
Particle Size:
0.3μm
1μm
2.5μm
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Particle Size
LSKMAG25C02 2 x 1mL 2.5μm
LSKMAG03C10 10 mL 0.3μm
LSKMAG1C10 10 mL 1μm
LSKMAG03C02 2 x 1mL 0.3μm
LSKMAG25C10 10 mL 2.5μm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. LSKMAG25C02 Supplier MilliporeSigma™ Supplier No. LSKMAG25CBX02
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For rapid and reproducible isolation of proteins

Custom affinity bead generation for binding to target ligand.

  • Advanced chemistry combined with high surface area provides more binding sites for proteins
  • Particles visible as they adhere to side of tube for quick and easy aspiration and complete buffer removal
  • Beads are immobilized in seconds
  • Increased kinetics of bead-protein binding enables shorter incubations
  • Significantly more affordable then competitive magnetic beads

Research Use Only

TRUSTED_SUSTAINABILITY

Specifications

Product Type Carboxy flexiBind Magnetic bead system
Quantity 2 x 1mL
Particle Size 2.5μm
Content And Storage 2° to 8°C
For Use With (Application) Immunoprecipitations, purifying Nucleic Acids, isolating cells/organelles, Protein-protein interactions
Matrix Magnetic Beads
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.