Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ PureProteome™ Protein A/G Mix Magnetic Bead System

Catalog No. p-4645018 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mL
2 x 1mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LSKMAGAG02 2 x 1mL
LSKAGAG10 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LSKMAGAG02 Supplier MilliporeSigma™ Supplier No. LSKMAGAG02

powerful system to isolate antibodies faster, easier, and with better reproducibly then ever before

The PureProteome Protein A/G Mix Magnetic Bead System provides researchers with the power of the combination of both Protein A and Protein G immunoglobulin binding affinities in one convenient magnetic bead product, resulting in greater antibody compatibility than either Protein A or Protein G alone.

  • Suitable for immunoprecipitation, crosslinking immunoprecipitation, antibody purification
  • Binding capacity: 2–3 mg of Rabbit IgG per mL of suspension
  • High capacity: Advanced chemistry combined with high surface area provides more binding sites for proteins
  • Consistent results with no sample loss: Particles visible as they adhere to side of tube for quick and easy aspiration and complete buffer removal
  • Fast processing time: Beads are immobilized in seconds; increased kinetics of bead-protein binding enables shorter incubations
  • Particle Size: 10μm

TRUSTED_SUSTAINABILITY

Specifications

Product Type Protein A/G mix Magnetic bead
Quantity 2 x 1mL
Particle Size 10μm
Content And Storage 2° to 8°C
For Use With (Application) Immunoprecipitation, cross-linking immunoprecipitation, Antibody purification
Matrix Magnetic Beads
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.