Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Q-Gard TL Polishing Packs for ELIX, RiOs™

Catalog No. 09729070 Shop All MilliporeSigma Products
Change view
Click to view available options
Polishing Pack; For ELIX, RiOs systems
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
09-729-070 Polishing Pack; For ELIX, RiOs systems
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-729-070 Supplier MilliporeSigma™ Supplier No. QGARDTL01
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For ELIX, RiOs (30 to 200) systems

Contains virgin ion-exchange resins for Elix
TRUSTED_SUSTAINABILITY

Specifications

Purification Method Ion Exchange, Depth Filtration
For Use With (Equipment) RiOs™ (30 to 200), Elix™ Clinical (35 to 70), Elix™ Golfstream Clinical, Large Elix™ (20 to 100)
Model Q-Gard TL
Type Polishing Pack
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.