Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Q-POD 2™ Ultrapure Water Remote Delivery Point
GSA_VA

Catalog No. ZMQSP0D02 Shop All MilliporeSigma Products
Change view
Click to view available options
Millipore Q-POD 2 remote dispenser
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
ZMQSP0D02 Millipore Q-POD 2 remote dispenser
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Catalog No. ZMQSP0D02 Supplier MilliporeSigma™ Supplier No. ZMQSP0D02

missing translation for 'validMainframeMsg'

This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.

For convenient access to ultrapure water

Connect the Q-POD 2™ remote dispenser to a Milli-Q Reference system or Milli-Q Direct, and ultrapure water is always within reach.

  • For polishing and delivery of Type 1 water
  • Product water resistivity: Ultrapure 18.2
  • Processes 1 to 200L per day
  • Flow rate up to 2L/min.

Polishing and delivery of Type 1 water

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Capacity (English) 0.2 to 52.8
Cartridge Type Millipak™ and BioPak™
Capacity (Metric) 1 to 200
Flow Rate 2
Product Water Type Ultrapure (Type 1)
Type Remote Dispenser
For Use With (Equipment) Millipore™ Q-POD™ Ultrapure™ Water Remote Delivery Point
Range (Conductivity) 0.056
Resistivity 18.2MΩ·cm
Height (English) 22.8 in.
Height (Metric) 58 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.