Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Qpak Ion Exchange Resin Cartridge

Catalog No. CP4ALLRES Shop All MilliporeSigma Products
Change view
Click to view available options
Qpak Ion Exchange Resin Cartridge
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
CP4ALLRES Qpak Ion Exchange Resin Cartridge
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CP4ALLRES Supplier MilliporeSigma™ Supplier No. CP4ALLRES

missing translation for 'validMainframeMsg'

With ion-exchange technology

Polishing cartridge with virgin ion-exchange resins for AFS™ 10/18/30/60 systems and AFS™ 25EDI/50EDI systems
TRUSTED_SUSTAINABILITY

Specifications

Material Resin
Purification Method Ion Exchange
For Use With (Application) Polishing
For Use With (Equipment) AFS™ 10/18/30/60 Systems and AFS 25EDI/50EDI Systems
Model QPAK ion exchange resin
Quantity 1/Pk.
Type Polishing Cartridge
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.