Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Quantum IX Cartridge

Catalog No. QTUMMPKIX Shop All MilliporeSigma Products
Change view
Click to view available options
Pore Size:
0.2 μm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pore Size
QTUMMPKIX 0.2 μm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. QTUMMPKIX Supplier MilliporeSigma™ Supplier No. QTUMMPKIX
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specific for removal of ions with point-of-use final filter.

Kit: Quantum™ IX Cartridge & Millipak™ 40 Filter
TRUSTED_SUSTAINABILITY

Specifications

Pore Size 0.2 μm
Purification Method Ion Exchange, Membrane Filter
Resin Type Ionex
For Use With (Equipment) Milli-Q™ academic, biocel, element, gradient, synthesis
Type Cartridge
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.