Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Reflectoquant™ Calcium Test Kit

Catalog No. p-4688198 Shop All MilliporeSigma Products
Change view
Click to view available options
Graduations:
2.5 to 45mg/L
5 to 125mg/L
Range:
2.5 to 45 mg/L
5 to 125 mg/L
Storage Requirements:
15°C to 25°C
2°C to 8°C
Test Method:
Glyoxalbishydroxyanil
Phthaleinkomplexone
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Graduations Range Storage Requirements Test Method
M1161250001 5 to 125mg/L 5 to 125 mg/L 2°C to 8°C Phthaleinkomplexone
M1169930001 2.5 to 45mg/L 2.5 to 45 mg/L 15°C to 25°C Glyoxalbishydroxyanil
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1161250001 Supplier MilliporeSigma™ Supplier No. 1.16125.0001
Only null left
Add to Cart
Add to Cart

Includes

Reagent, if needed

Used in reflectometric test

  • For testing boiling and cooling water, drinking water, beer
  • Indicates possible presence of calcium deposits that can affect operation of machinery

Range: 5 to 125mg/L

Order Info

Pack of 50 strips

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Reflectometric
Graduations 5 to 125mg/L
Quantity 50 Tests
Range 5 to 125 mg/L
Storage Requirements 2°C to 8°C
Test Method Phthaleinkomplexone
Type Calcium Test Strip
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.