Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Reflectoquant™ Glucose Test Strips

Catalog No. M1167200001 Shop All MilliporeSigma Products
Change view
Click to view available options
No. of Tests:
50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Tests
M1167200001 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1167200001 Supplier MilliporeSigma™ Supplier No. 1.16720.0001
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Tube containing 50 test strips, 1 bar-code strip.

To determine the presence or absence of residues of glucose

  • Enzymatic reaction method
  • Barcode calibration for reliable, quantitative results
  • Low analysis costs

Reclose the tube containing the test strips immediately after use.

For food and breverage analysis

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Determination of glucose in beverages, food/preserves and fermentation solutions
For Use With (Equipment) Reflectoquant™ RQflex™ 10m reflectometer analysis system
Graduations 1 to 100mg/L
No. of Tests 50
Range 1 to 100mg/L
Storage Requirements 2°C to 8°C
Test Method Enzymatic Reaction
Type Test Strip
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.