Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Reflectoquant™ Peracetic Acid Test Strips

Catalog No. p-4626419 Shop All MilliporeSigma Products
Change view
Click to view available options
Range:
1 to 22.5mg/L
75 to 400mg/L
Graduations:
1 to 22.5mg/L
75 to 400mg/L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Range Graduations
M1169750001 1 to 22.5mg/L 1 to 22.5mg/L
M1169760001 75 to 400mg/L 75 to 400mg/L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1169750001 Supplier MilliporeSigma™ Supplier No. 1.16975.0001
Only null left
Add to Cart
Add to Cart

Includes

Tube containing 50 strips

For determination of inorganic peracetic acid in aqueous solutions

  • Redox reaction method
  • Barcode calibration for reliable, quantitative results
  • Low analysis costs

Reclose the tube containing the test strips immediately after use.

For food and water analysis

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Food and water analysis, ideal for determining when hydrogen peroxide is present in a sample
For Use With (Equipment) RQflex™ 10 and RQflex™ Plus 10 reflectometers
Graduations 1 to 22.5mg/L
No. of Tests 50
Range 1 to 22.5mg/L
Storage Requirements 2°C to 8°C
Test Method Redox Reaction
Type Test Strip
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.