Learn More
MilliporeSigma™ Reflectoquant™ Sucrose Test Strips
Catalog No. M1161410001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 Tests
Catalog No. Quantity
M1161410001 50 Tests
1 options
Catalog No. M1161410001 Supplier MilliporeSigma™ Supplier No. 1.16141.0001
Add to Cart
Add to Cart
Includes
Reagent, if needed
Used in reflectometric test
  • For the determination of sucrose in beverages such as fruit/vegetable juices and refreshment drinks after appropriate sample pretreatment or dilution
Range: 0.25 to 2.50g/L
Order Info
Pack of 50 strips
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Used for determination of sucrose in beverages, eg. fruit/vegetable juice and refreshment drinks after appropriate sample pretreatment or dilution
Graduations 0.25 to 2.5g/L
Includes Barcode strip, Sa-1 reagent bottle, 12mL graduated plastic syringe, 1mL graduated plastic syringe, 1 test vessel with stopper
Quantity 50 Tests
Storage Requirements 2°C to 8°C
Test Method Enzymatic reaction
Type Sucrose Test Strip