Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Reflectoquant™ Sulfite Test Strips

Catalog No. M1169870001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1169870001 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1169870001 Supplier MilliporeSigma™ Supplier No. 1.16987.0001
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Reagent, if needed

Used in reflectometric test

  • For determining sulfite ions
  • Ions form a red compound in reaction with reagents
  • Use in wasterwater, boiler water, boiler feed water and food after appropriate sample pretreatment

Range: 10 to 200mg/L as sulfite

Order Info

Pack of 50 strips

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Used for the determination of sulfite ions
For Use With (Equipment) Reflectometric
Graduations 10 to 200mg/L
Quantity 50 Tests
Range 10 to 200 mg/L
Storage Requirements 2°C to 8°C
Test Method Nitroprusside, Zn-hexacyanoferrate
Type Sulfite Test Strip
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.