Learn More
MilliporeSigma™ Reflectoquant™ Total Sugar Test Strips
Catalog No. M1161360001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 Tests
Catalog No. Quantity
M1161360001 50 Tests
1 options
Catalog No. M1161360001 Supplier MilliporeSigma™ Supplier No. 1.16136.0001
Add to Cart
Add to Cart
Includes
Reagent, if needed
Used in reflectometric test
  • Monitor glucose and fructose in food, such as potatoe and fruit juices
  • Optimize winemaking process
Range: 65 to 650mg/L
Order Info
Pack of 50 tests
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) With the total sugar test you can monitor glucose and fructose in food, such as potatoes, and in fruit juices and also optimize your winemaking process
Graduations 65 to 650mg/L
Includes 1 barcode strip, 1 bottle of reagent TS-1, 1 graduated 1mL plastic syringe, 1 graduated 12mL plastic syringe, 1 test vessel with stopper
Quantity 50 Tests
Range 65 to 650 mg/L
Storage Requirements 2°C to 8°C
Test Method Reflectometric
Type Reflectoquant™ Total Sugar Test Strip