Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Upstate™ REST, Rabbbit Polyclonal, ChIP Validated Antibody and Primer Set

Catalog No. 1710456MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 Assays
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
17-104-56MI 25 Assays Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-104-56MI Supplier MilliporeSigma™ Supplier No. 1710456
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Specifically detects REST Clone: in Human, Mouse, Rat samples, and it is validated for ChIP, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen REST
Applications ChIP Assay, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Anti-REST (Rabbit Polyclonal). One vial containing 100μL 1.0mg/mL purified rabbit polyclonal in buffer containing 0.014M phosphate buffer, pH 7.6, 0.175M NaCl, 0.07% sodium azide, and 30% glycerol. Normal Rabbit IgG. One vial containing 125μg Rabbit IgG in 125μL storage buffer containing 0.05% sodium azide. ChIP Primers, human Snap25. One vial containing 75μL of 5μM of each primer specific for human Snap25 promoter. FOR: AGA CTC CTT TGC AGA CAA TTT CCT; REV: CAA CAC AGA AGA TTT CCA CAA GTA GAC
Gene Accession No. Q13127
Gene Symbols NRSF, XBR
Host Species Rabbit
Immunogen The REST purified antibody is made against a GST fusion protein corresponding to amino acids 801-1097 of human REST.
Quantity 25 Assays
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_005603
Test Specificity Recognizes REST, MW 125kDa predicted, 200kDa observed.
Includes This ChIPAb+ REST -ChIP Validated Antibody and Primer Set conveniently includes the antibody and the specific control PCR primers.
Target Species Human, Mouse, Rat
Content And Storage -20°C for up to 12 months from date of receipt; Upon first thaw and prior to removing the cap centrifuge the vial and gently mix the solution; Aliquot into microcentrifuge tubes and store at −20°C; Avoid repeated freeze/thaw cycles
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.