Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ RiOs™ 30/50/100 Water Purification Systems

Catalog No. ZR0S60050 Shop All MilliporeSigma Products
Change view
Click to view available options
Model: RiOs 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
ZR0S60050 Model: RiOs 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ZR0S60050 Supplier MilliporeSigma™ Supplier No. ZR0S60050

missing translation for 'validMainframeMsg'

Requires

  • Requires 8mm tubing for product and Elix module reject water
  • 1/2 in. tubing for RO reject water
  • >11/3 gal./min. (5L/min.) drain

For ensuring quality, the Millipore RiOs™ 30/50/100 Water Purification System can serve a single lab, suite of labs, or an entire laboratory building, depending on model.

Combine water purification, storage, distribution and control in a single unit. Provide hundreds to thousands of L/day of Type III laboratory grade water from tap.

Key Features

  • Utilize a three-step water treatment process
  • Provide total control of all functions, operating parameters and standard accessory components within the RiOs system itself and within the external pure water distribution loop
  • Do not usually require an external control box

Pretreatment Features

  • Progard™ L pack removes particles (0.5μm filter), free chlorine, and colloids (activated carbon filter) from tap water to protect the system
  • Anti-scaling agent protects the reverse osmosis membrane in hard water areas
  • Bactericidal carbon prevents unwanted bacteria growth
  • Pretreatment packs are tailored to the feedwater source
Advanced Reverse Osmosis (RO) Features
  • Removes 95 to 99% of inorganic ions and 99% of all dissolved organics (MW 200 Daltons), microorganisms and particles
  • Recycles part of the RO reject water back to the RO membrane feedwater stream
  • Allows water recovery to be adjusted up to 70% to optimize water consumption, depending on the feedwater quality and the pretreatment sequence used
  • Maintains constant flowrate from 7 to 30°C
Ultraviolet Sanitization Features
  • Uses a 254nm UV lamp
  • Leads to a log reduction value (LRV) of four in the bacteria count of the water, irrespective of the system's nominal flowrate
System Operation Features
  • After each purification step, relevant parameters are checked by the system
  • Water quality monitoring within the pure water distribution loop (resistivity) can also be checked by the system
  • Resistivity test is designed to meet the requirements of USP (645) suitability test
  • Water quality data is recorded by the system in real time for full traceability
  • Solenoid valve feed: 1/2 NPT
  • RS-232 data port connection

  • Produces water ideal for general rinsing, bacteria-sensitive applications, or as feedwater for humidifiers, autoclaves and washing machines
  • Also suitable as a source of feedwater for Milli-Q™ and Super-Q™ ultrapure water systems

†Includes plumbing connections.

TRUSTED_SUSTAINABILITY

Specifications

Conductivity <2000μS/cm
Contaminant Rejection (Microbiological) Less than10cfu/mL
Contaminant Rejection (Organic) Greater than99.9%
Contaminant Rejection (Particulate) Greater than99.9%
Daily Output 1000 L
Feedwater Source Potable Tap Water
Flow Rate Less than 5
Product Water Type Type 3
Type Water Purification System
Feedwater Pressure 2 to 6 bar
Filtration Area (Metric) 0.5 μm
Model RiOs 50
Modules Elix™
pH 4 to 10
Temperature Range (English) 44.6°F to 86°F
Temperature Range (Metric) 7°C to 30°C
Dimensions (L x W x H) 17.37 x 26.1 x 28.9 in. (44 x 66.2 x 73.4 cm)
Length (English) 17.37 in.
Length (Metric) 44 cm
Width (English) 26.1 in.
Width (Metric) 66.2 cm
Height (English) 28.9 in.
Height (Metric) 73.4 cm
Depth (English) 17.4 in.
Depth (Metric) 44.1 cm
Weight (English) 75 lb.
Weight (Metric) 34 kg
Electrical Requirements 110 V 60 Hz
Voltage 110 V
Frequency 60 Hz
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.