Learn More
MilliporeSigma™ γ-Secretase Inhibitor XXIII, Aβ42-Selective, Calbiochem, Protein
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Preparation Method | DMSO (25mg/mL) |
| Purification Method | HPLC |
| Quantity | 5 mg |
| Immunogen | γ-Secretase Inhibitor XXIII, Aβ42-Selective |
| Storage Requirements | Following reconstitution, aliquot and freeze (−20°C). Stock solutions are stable for up to 2 months at −20°C. |
| Regulatory Status | RUO |
| Recombinant | Native |
| Form | Solid |
| Purity or Quality Grade | ≥95% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.