Learn More
MilliporeSigma™ SeQuant™ SAMS™ and CARS™ Suppressor System: Installation Kit
Catalog No. M1506190001 Shop All MilliporeSigma Products
Change view
Click to view available options
For Use With (Equipment):
CARS Suppressor
Catalog No. For Use With (Equipment)
M1506190001 CARS Suppressor
1 options
Catalog No. M1506190001 Supplier MilliporeSigma™ Supplier No. 1.50619.0001
tubing (5 m) and fittings (6 ea) for CARS™ system SeQuant™
SeQuant™ CARS™ (Continuous Anion Regeneration System) with SAMS™ (SeQuant™ Anion Membrane Suppressor) will enhance and simplify your anion chromatography analysis. The continuous chemical suppression of the eluent reduces the background and increases the signal for the analyte anions.
This tubing kit is required for the installation of SAMS™ and is also included in the CARS™ system.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) CARS Suppressor
Type Installation kit