Learn More
MilliporeSigma™ SeQuant™ SAMS™ and CARS™ Suppressor System: Pressure Relief Valve
Catalog No. M1506180001 Shop All MilliporeSigma Products
Change view
Click to view available options
For Use With (Equipment):
SeQuant SAMS Suppressors
Catalog No. For Use With (Equipment)
M1506180001 SeQuant SAMS Suppressors
1 options
Catalog No. M1506180001 Supplier MilliporeSigma™ Supplier No. 1.50618.0001
back-pressure guard valve 100 psi for SAMS™ suppressors SeQuant™
SeQuant™ CARS™ (Continuous Anion Regeneration System) with SAMS™ (SeQuant™ Anion Membrane Suppressor) will enhance and simplify your anion chromatography analysis. The continuous chemical suppression of the eluent reduces the background and increases the signal for the analyte anions.
This pressure relief valve protects your SAMS™ suppressor from breaking at high backpressures and is recommended for new installations.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) SeQuant SAMS Suppressors
Type Pressure Relief Valve