Learn More
MilliporeSigma™ SeQuant™ SAMS™ and CARS™ Suppressor System: Replacement Cartridges
Catalog No. M1506140001 Shop All MilliporeSigma Products
Change view
Click to view available options
Type:
Large Replacement cartridge
Small Replacement cartridge
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
M1506140001 Large Replacement cartridge
M1506130001 Small Replacement cartridge
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1506140001 Supplier MilliporeSigma™ Supplier No. 1.50614.0001
For use with CARS™ system SeQuant™
SeQuant™ CARS™ (Continuous Anion Regeneration System) with SAMS™ (SeQuant™ Anion Membrane Suppressor) will enhance and simplify your anion chromatography analysis. The continuous chemical suppression of the eluent reduces the background and increases the signal for the analyte anions.
This cartridge replaces the one supplied with the CARS™ system when it is depleted.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) SeQuant SAMS and CARS Suppressor System
Type Large Replacement cartridge