Learn More
MilliporeSigma™ SAMS Standard Suppressor
Catalog No. M1506090001 Shop All MilliporeSigma Products
Change view
Click to view available options
For Use With (Equipment):
SAMS and CARS Suppressor System
Catalog No. For Use With (Equipment)
M1506090001 SAMS and CARS Suppressor System
1 options
Catalog No. M1506090001 Supplier MilliporeSigma™ Supplier No. 1.50609.0001
Includes
One suppressor 100cm membrane, 10-32 fittings, 1 fitting kit with 10-32 UNF fittings for 1/16 in. tubing, 1 syringe
For analysis in anion ion chromatography.
  • With chemically regenerated membrane (100cm) for anion chromatography
    • Conductivity: ≤3μS/cm
    • Pressure: ≤300 kPa
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) SAMS and CARS Suppressor System
Type Standard Suppressor
Includes One suppressor 100 cm membrane, 10-32 fittings, 1 fitting kit with 10-32 UNF fittings for 1/16 in. tubing, 1 syringe