Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Serotonin 3 (5-HT3) Receptor (444-457), Rabbbit Polyclonal, ChIP Validated Antibody and Primer Set

Catalog No. PC3471SET Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Set
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
PC3471SET 1 Set Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PC3471SET Supplier MilliporeSigma™ Supplier No. PC3471SET

Rabbit Polyclonal Antibody

Specifically detects Serotonin 3 (5-HT3) Receptor (444-457) Clone: in Mouse, Rat samples, and it is validated for Immunohistochemistry (Frozen)

The monoamine serotonin (5-hydroxytryptamine) mediates a variety of physiological actions by binding to a number of distinct receptors, pharmacologically classified into several major groups which differ in tissue and cellular localization, affinity for serotonin, and second messenger pathways. The majority of these receptors stimulate a GTP-binding protein upon agonist stimulation and couple to adenylate cyclase or phospholipase C. In contrast, the 5-HT3 receptor is a ligand-gated ion channel that mediates a fast excitatory response. This receptor is a member of the superfamily of ligand-gated ion channels related to the nicotinic, GABAA and NMDA receptors. Although the 5-HT3 receptor was initially identified from the neuroblastoma cell line NCB-20, radioligand binding studies and in situ hybridization studies have localized the 5-HT3 receptor to a unique distribution of neurons, supporting possible roles for the receptor in cognition and sensory input. Recent immunohistochemical analysis of the rat brain revealed a broad distribution of 5-HT3 immunoreactivity, primarily including interneurons and motor neurons.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Serotonin 3 (5-HT3) Receptor (444-457)
Applications Immunohistochemistry (Frozen)
Classification Polyclonal
Conjugate Unconjugated
Formulation 25μg antibody in 0.05 M sodium phosphate buffer, 0.2% gelatin and 25μg lyophilized control peptide.
Host Species Rabbit
Immunogen a synthetic peptide (SLEKRDEMREVARD) corresponding to amino acids 444-457 of rat serotonin (5-HT3) receptor, conjugated to KLH
Quantity 1 Set
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the serotonin 3 (5-HT3) receptor in mouse and rat brain tissue. Supplied with a control peptide.
Includes 25μg antibody in 0.05M sodium phosphate buffer, 0.2% gelatin and 25μg lyophilized control peptide.
Target Species Mouse, Rat
Content And Storage 2°C to 8°C
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.