Learn More
MilliporeSigma™ Silica Gel 60G TLC Plates, Glass Backed
Catalog No. M1003840001 Shop All MilliporeSigma Products
Change view
Click to view available options
Type:
TLC Silica Gel 60 G
TLC Silica Gel 60 G F254
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
M1003840001 TLC Silica Gel 60 G
M1003900001 TLC Silica Gel 60 G F254
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1003840001 Supplier MilliporeSigma™ Supplier No. 1.00384.0001
Add to Cart
Add to Cart
These new TLC silica gel 60G plates are recommended for customers in QA/QC labs using older Ph Eur monograph methods, which require TLC plates with gypsum binder.
  • 20 x 20cm
  • Provided in packs of 25
  • For thin layer chromatography applications
TRUSTED_SUSTAINABILITY
Specifications
Quantity 25/Pk.
Type TLC Silica Gel 60 G
Dimensions (L x W) 20 x 20 cm