Learn More
MilliporeSigma™ Simplicity™ Ultrapure Water System: Simpak 2 Kit
GSA_VA
Catalog No. SIMPAKKD2 Shop All MilliporeSigma Products
Change view
Click to view available options
Feedwater Source:
Deionized
Catalog No. Feedwater Source
SIMPAKKD2 Deionized
1 options
Catalog No. SIMPAKKD2 Supplier MilliporeSigma™ Supplier No. SIMPAKKD2
Add to Cart
Add to Cart
For use with Simplicity and Simplicity 185 Ultrapure Water Systems
  • For pretreated deionized water
  • Includes one each Purification Pack, SimFilter, and 0.05μm Reservoir Vent Filter
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Feedwater Source Deionized
Pore Size 0.05 μm
For Use With (Equipment) Simplicity™ 185 Ultrapure Water System
Includes One Simpak two purification pack, one reservoir vent filter and one 0.05μm Simfilter final filter
Model SimPak 2
Type Cartridge Kit