Learn More
MilliporeSigma™ SmartPak™ Purification Cartridges
Catalog No. SPR0LS001US Shop All MilliporeSigma Products
Change view
Click to view available options
For Use With (Equipment):
RiOs™ 3 and 5 systems
Model:
SmartPak™
Catalog No. For Use With (Equipment) Model
SPR0LS001US RiOs™ 3 and 5 systems SmartPak™
1 options
Catalog No. SPR0LS001US Supplier MilliporeSigma™ Supplier No. SPR0LS001US
Use for pretreatment of tap water
  • Filtration tecnology used: Activated carbon, reverse osmosis, and ion-exchange
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) RiOs™ 3 and 5 systems
Model SmartPak™
Type Low Temperature RO Purification Pack