Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Upstate™ SMRTα, Mouse monoclonal, ChIP Validated Antibody and Primer Set

Catalog No. 1710057MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 Assays
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
17-100-57MI 25 Assays Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-100-57MI Supplier MilliporeSigma™ Supplier No. 1710057
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects SMRTα Clone: in Human, Mouse, Rat samples, and it is validated for ChIP, Electromobility Shift Assay, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blotting

The Silencing mediator of retinoic acid & thyroid hormone receptor protein, commonly called the SMRT protein mediates the transcriptional repression of some nuclear receptors by promoting chromatin condensation, thus preventing access of the basal transcription machinery. Consistent with this activity, this protein is known to form a large corepressor complex containing SIN3A/B and histone deacetylases HDAC1 and HDAC2. SMRT is also a component of the N-Cor repressor (nuclear receptor corepressor), a multi-subunit complex minimally composed of NCOR1, NCOR2, HDAC3, TBL1X, TBL1R, CORO2A and GPS2. SMRT and nuclear receptor corepressor N-CoR are related transcriptional corepressors which contain two distinct domains capable of interacting with unliganded nuclear receptors to repress their basal transcriptional activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMRTα
Applications ChIP Assay, Electromobility Shift Assay, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Conjugate Unconjugated
Formulation Anti-SMRT (mouse monoclonal). One vial containing 50μL of unpurified mouse monoclonal ascites with 0.05% sodium azide. Negative Ascites (mouse). One vial containing 50μL of mouse ascites with 0.05% sodium azide. ChIP Primers, IκBα promoter. One vial containing 75μL of 5μM of each primer specific for human IĸBα promoter. FOR: GAC GAC CCC AAT TCA AAT CG; REV: TCA GGC TCG GGG AAT TTC C
Gene Accession No. Q9Y618
Gene Symbols NCOR2, CTG26, N-CoR2, SMAP270, SMRT, SMRTE, SMRTE-tau, TNRC14, TRAC, TRAC-1, TRAC1
Host Species Mouse
Immunogen Recombinant protein corresponding to human SMRT.
Purification Method Unpurified
Quantity 25 Assays
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_001070729.1
Test Specificity Recognizes SMRT.
Includes The ChIPAb+ SMRT set includes the SMRT antibody, a negative control mouse ascites and qPCR primers which amplify a 299bp region of human IκBα promoter.
Target Species Human, Mouse, Rat
Content And Storage -20°C for one year from date of shipment; Upon first thaw and prior to removing the cap centrifuge the vial and gently mix the solution; Aliquot into microcentrifuge tubes and store at −20°C; Avoid repeated freeze/thaw cycles
Form Ascites
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.