Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Upstate™ Sox-2, clone 6F1.2α, 6F1.2, ChIP Validated Antibody and Primer Set

Catalog No. 17656MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 Assays
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
17-656-MI 25 Assays Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-656-MI Supplier MilliporeSigma™ Supplier No. 17656
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Specifically detects Sox-2, clone 6F1.2α Clone: 6F1.2 in Human, Mouse samples, and it is validated for Chromatin Immunoprecipitation, Dot Blot, Immunocytochemistry, Western Blotting

All ChIPAb+ antibodies are individually validated for chromatin precipitation, every lot, every time. Each ChIPAb+ antibody set includes control primers (tested every lot by qPCR) to biologically validate your IP results in a locus-specific context. The qPCR protocol and primer sequences are provided, allowing researchers to validate ChIP protocols when using our antibody in their chromatin context. Each set also includes a negative control antibody to ensure specificity of the ChIP reaction. The ChIPAb+ Sox-2, clone 6F1.2 antibody/primer set includes the Sox-2, clone 6F1.2 antibody, a negative control antibody (purified mouse IgG), and qPCR primers which amplify a 95bp region within the promoter of the human nanog gene. The Sox-2, clone 6F1.2 antibody and negative control antibodies are supplied in a scalable 'per ChIP' reaction size and can be used to functionally validate the precipitation of Sox-2-associated chromatin. Sox-2 plays a key role in embryonic stem cell development, and has been shown to help stem cells maintain pluripotent potential. It has also been shown to be associated with uncommitted dividing stem cell and precursor cells of the developing CNS.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Sox-2, clone 6F1.2α
Applications ChIP Assay, Dot Blot, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 6F1.2
Conjugate Unconjugated
Formulation Anti-Sox-2, clone 6F1.2 (mouse monoclonal IgG). One vial containing 100μg of protein A purified antibody in 140μL of 0.02M phosphate buffer pH 7.6 with 0.25M NaCl containing 0.1% sodium azide, and 30% glycerol. ChIP Primers, nanog promoter. One vial containing 75μL of 5μM of each primer specific for the promoter region of human nanog. FOR: GTT CTG TTG CTC GGT TTT CT; REV: TCC CGT CTA CCA GTC TCA CC
Gene Accession No. P48431
Gene Symbols SOX2; ANOP3; MGC2413; MCOPS3
Host Species Mouse
Immunogen Recombinant GST Fusion protein.
Quantity 25 Assays
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_003097
Test Specificity Recognizes Sox-2, MW: ∽34kDa.
Includes This ChIPAb+ Sox-2, clone 6F1.2 -ChIP Validated Antibody and Primer Set conveniently includes the antibody and the specific control PCR primers.
Target Species Human, Mouse
Content And Storage -20°C for one year from date of shipment; Upon first thaw and prior to removing the cap centrifuge the vial and gently mix the solution; Aliquot into microcentrifuge tubes and store at −20°C; Avoid repeated freeze/thaw cycles
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.