Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Upstate™ SOX-2α, Mouse monoclonal, ChIP Validated Antibody and Primer Set

Catalog No. 1710256MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 Assays
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
17-102-56MI 25 Assays Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-102-56MI Supplier MilliporeSigma™ Supplier No. 1710256

Mouse Monoclonal Antibody

Specifically detects SOX-2α Clone: in Human, Mouse samples, and it is validated for Chromatin Immunoprecipitation, Immunocytochemistry, Western Blotting

SOX 2 is also known as SRY related HMG BOX gene 2. All SOX proteins have a single HMG box and bind linear DNA in a sequence-specific manner, resulting in the bending of DNA through large angles. Bending causes the DNA helix to open for some distance, which may affect binding and interactions of other transcription factors. SOX1, SOX2 and SOX3 show the closest homology to SRY. They share maximum homology within the HMG domain and are expressed mainly in the developing nervous system of the mouse. These genes share significant homology outside the HMG box also and are highly conserved throughout their evolution. Sox-2 plays a key role in embryonic stem cell development, and has been shown to help stem cells maintain a pluripotent potential. It has also been shown to be associated with uncommitted dividing stem cell and precursor cells of the developing CNS.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SOX-2α
Applications ChIP Assay, Immunocytochemistry, Western Blot
Classification Monoclonal
Conjugate Unconjugated
Formulation Anti-SOX-2 (mouse monoclonal). One vial containing 75μg of protein G purified monoclonal in buffer containing 0.1M Tris-glycine (pH 7.4), 150mM NaCl, pH 7.4, 0.05% sodium azide before the addition of glycerol to 30%. Concentration: 0.7mg/mL. Normal Mouse IgG. One vial containing 125μg purified mouse IgG in 125μL storage buffer containing 0.1% sodium azide. ChIP Primers, nanog promoter. One vial containing 75μL of 5μM of each primer specific for the promoter region of human nanog. FOR: GTT CTG TTG CTC GGT TTT CT; REV: TCC CGT CTA CCA GTC TCA CC
Gene Accession No. P48431
Gene Symbols SOX2
Host Species Mouse
Immunogen Recombinant protein corresponding to human SOX-2.
Purification Method Protein G purified
Quantity 25 Assays
Regulatory Status RUO
Research Discipline Stem Cell Research
Primary or Secondary Primary
Gene ID (Entrez) NP_003097
Includes This ChIPAb+ SOX-2 -ChIP Validated Antibody and Primer Set conveniently includes the antibody and the specific control PCR primers.
Target Species Human, Mouse
Content And Storage -20°C for one year from date of shipment; Upon first thaw and prior to removing the cap centrifuge the vial and gently mix the solution; Aliquot into microcentrifuge tubes and store at −20°C; Avoid repeated freeze/thaw cycles
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.