Learn More
MilliporeSigma™ Special Indicator pH Strip, 5.2 to 7.2
Catalog No. M1096320001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 Strips
Catalog No. Quantity
M1096320001 100 Strips
1 options
Catalog No. M1096320001 Supplier MilliporeSigma™ Supplier No. 1.09632.0001
Add to Cart
Add to Cart
for pH measurements in meat non-bleeding pH 5.2 - 5.6 - 6.0 - 6.4 - 6.8 - 7.2 MColorpHast™
Special indicator test strip for the pH measurement directly in meat. No contamination of the measurement site thanks to the covalent binding of the indicator dyes.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) pH measurement directly in meat
Quantity 100 Strips
Type Special Indicator pH Strip