Learn More
Description
Specifications
Specifications
| Concentration | Concentration on Label |
| Preparation Method | 5mg/mL in 50mM potassium phosphate, 100μM EDTA, pH 7.8, yields a clear pale blue solution. |
| Quantity | 50 kU |
| Immunogen | Superoxide Dismutase |
| Storage Requirements | Following reconstitution, aliquot and freeze (−20°C). |
| Regulatory Status | RUO |
| Species | Bovine |
| Recombinant | Native |
| Form | Lyophilized |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.