Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Upstate™ SUZ12α, Mouse monoclonal, ChIP Validated Antibody and Primer Set

Catalog No. 17661MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 Assays
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
17-661-MI 25 Assays Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-661-MI Supplier MilliporeSigma™ Supplier No. 17661
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Specifically detects SUZ12α Clone: in Human, Mouse, Rat, Vertebrate samples, and it is validated for ChIP, Dot Blot, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen SUZ12α
Applications ChIP Assay, Dot Blot, Western Blot
Classification Monoclonal
Conjugate Unconjugated
Formulation Anti-SUZ12 (mouse monoclonal IgG). One vial containing 50μg of protein A purified antibody in PBS containing 0.05% sodium azide. May contain 30% glycerol (see certificate of analysis). Normal Mouse IgG. Two vials containing 25μg of purified mouse IgG in 25μL of storage buffer containing 0.1% sodium azide. ChIP Primers, HoxA2 upstream. One vial containing 75μL of 5μM of each primer specific for the promoter region of human HoxA2. FOR: AGG AAA GAT TTT GGT TGG GAA G; REV: AAA AAG AGG GAA AGG GAC AGA C
Gene Accession No. Q15022
Gene Symbols SUZ12; JJAZ1; ChET9
Host Species Mouse
Immunogen SUZ12, purified antibody is made against human SUZ12.
Quantity 25 Assays
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_056170
Test Specificity Recognizes SUZ12, Mr 95kDa.
Includes This ChIPAb+ SUZ12 -ChIP Validated Antibody and Primer Set conveniently includes the antibody and the specific control PCR primers.
Target Species Human, Mouse, Rat, Vertebrates
Content And Storage -20°C for one year from date of shipment; Upon first thaw and prior to removing the cap centrifuge the vial and gently mix the solution; Aliquot into microcentrifuge tubes and store at −20°C; Avoid repeated freeze/thaw cycles
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.