Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Synergy™ Ultrapure Water Purification System
GSA_VA

Catalog No. SYNSVR0US Shop All MilliporeSigma Products
Change view
Click to view available options
Total Organic Carbon:
<5 ppb
Includes:
Remote Dispenser, 185nm Photooxidation UV Lamp
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Total Organic Carbon Includes
SYNSVR0US <5 ppb Remote Dispenser, 185nm Photooxidation UV Lamp
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SYNSVR0US Supplier MilliporeSigma™ Supplier No. SYNSVR0US

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

Obtain Type 1 reagent grade water with a flow rate above 1L/min when using the Millipore Synergy™ Ultrapure Water Purification System.

Synergy systems are designed for laboratories that already have access to purified water and need a point-of-use system to supply high quality ultrapure water.

  • Produce Type 1 water (18.2 MOhm. cm at 25°C ultrapure water) on demand from pretreated water (reverse osmosis or demineralized water)
  • Maintenance reduced to a single cartridge change once or twice a year, depending on the volume produced and the quality of the pure water fee
  • Designed for users needing between 2 and 100 liters of ultrapure water per day
  • Quality suitable for HPLC and other spectrophotometric/spectroscopic analytical techniques for biological, biochemical, and cell culture applications
  • Dispensing adapted to different size laboratory glassware
  • Time-saving fixed-volume dispense
  • Compact design, along with the versatile Remote dispenser, ensure easy installation anywhere in your laboratory

  • Dimensions: 15D x 21.3W x 11.4 in. H (38 x 54 x 29cm)
  • Electrical feed voltage: 100-250V ±10%
  • Electrical feed frequency:50-60Hz ±10%

  • Synergy UV System: For applications requiring very low organic contaminant levels,it incorporates a dual wavelength UV lamp to produce water with <5ppb TOC (when feed water TOC is below 50ppb), making it suitable for HPLC, GC, ILC and TOC analyses

LC buffer preparation, HPLC isocratic and gradient solution preparation, Sample dilution, Buffer reagents for molecular biology and cell culture media preparation, Preparation of chemical solutions used with titrators, spectrophotometers and electrophoresis systems

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Feedwater Source Pretreated
Flow Rate 1 L/min.
Product Water Type Type 1
Resistivity 18.2 Mohm.cm product water
Total Organic Carbon <5 ppb
Feedwater Pressure 4.3 psi (0.3 bar)
For Use With (Application) Designed for laboratories that already have access to purified water and need a point-of-use system to supply high quality ultrapure water
Includes Remote Dispenser, 185nm Photooxidation UV Lamp
Width (English) 11.4 in.
Width (Metric) 29 cm
Height (English) 21.3 in.
Height (Metric) 54 cm
Depth (English) 15 in.
Depth (Metric) 38 cm
Electrical Requirements 100/240 V 50/60 Hz
Voltage 100/240 V
Frequency 50/60 Hz
Product Type UV Pure to Pure and Ultrapure Purification System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.