Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ α-Synuclein A53T, Human, Recombinant Protein, E. coli

Catalog No. 575003200UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
57-500-3200UG 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 57-500-3200UG Supplier MilliporeSigma™ Supplier No. 575003200UG
Only null left
Add to Cart
Add to Cart
Recombinant, human α-synuclein containing an A53T mutation and expressed in E. coli. A point mutant (A53T) of α-synuclein gene that has been linked to autosomal dominant early-onset Parkinson's disease (PD).
TRUSTED_SUSTAINABILITY

Specifications

Concentration Concentration on Label
Formulation In 100mM NaCl, 20mM Tris-HCl, pH 7.5.
Purification Method SDS-PAGE
Quantity 200 μg
Source E. coli
Immunogen α-Synuclein A53T
Storage Requirements Following initial thaw, aliquot and freeze (−20°C).
Regulatory Status RUO
Species Human
Recombinant Recombinant
Form Liquid
Purity or Quality Grade ≥95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.