Learn More
MilliporeSigma™ T7Select 1-2b DNA Vector
Catalog No. 700433 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
70-043-3 10 μg
1 options
Catalog No. 70-043-3 Supplier MilliporeSigma™ Supplier No. 700433
Add to Cart
Add to Cart
The T7Select™1-2a-c vectors are protein display vectors based on bacteriophage T7. The vector displays an average of 0.1–1 copies of peptides and proteins up to 900 aa in size on the surface of the T7 capsid. Target sequences are fused to the C-terminus of the 10B capsid protein near amino acid 348.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at 2° to 8°C.
Quantity 10 μg