Learn More
MilliporeSigma™ T7Select 10-3b DNA Vector
Catalog No. 705483 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
70-548-3 10 μg
1 options
Catalog No. 70-548-3 Supplier MilliporeSigma™ Supplier No. 705483
Add to Cart
Add to Cart
The T7Select™10-3b vector is a protein display vector based on bacteriophage T7. The vector displays an average of 5–15 copies of peptides and proteins up to 1200 aa in size on the surface of the T7 capsid. Target sequences are fused to the C-terminus of the 10B capsid protein near amino acid 348.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 10 μg