Learn More
MilliporeSigma™ T7Select 415-1 DNA Vector
Catalog No. 700403 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
70-040-3 10 μg
1 options
Catalog No. 70-040-3 Supplier MilliporeSigma™ Supplier No. 700403
Add to Cart
Add to Cart
The T7Select™ 415-1b vector is a protein display vector based on bacteriophage T7. The vector displays 415 copies of peptides up to 50 aa in size on the surface of the T7 capsid. Target sequences are fused to the C-terminus of the 10B capsid protein near amino acid 348. When T7Select415 recombinants are grown on normal T7 hosts such as BL21, the capsid shell is composed entirely of the fusion protein. The linear genome of T7Select415-1b is 37,314 bp in size.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at 2° to 8°C.
Quantity 10 μg