Learn More
MilliporeSigma™ T7SelectUP Primer
Catalog No. 700053 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 pmol
Catalog No. Quantity
70-005-3 500 pmol
1 options
Catalog No. 70-005-3 Supplier MilliporeSigma™ Supplier No. 700053
Add to Cart
Add to Cart
For amplification and sequencing of T7Select recombinants, and are compatible with all T7Select vectors
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at -20°C. Avoid freeze/thaw cycles.
Quantity 500 pmol