Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Upstate™ TATA Binding Protein (TBP)α, Mouse monoclonal, ChIP Validated Antibody and Primer Set

Catalog No. 1710098MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 Assays
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
17-100-98MI 25 Assays Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-100-98MI Supplier MilliporeSigma™ Supplier No. 1710098
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Specifically detects TATA Binding Protein (TBP)α Clone: in Chicken, Fish, Human, Mouse, Xenopus samples, and it is validated for ChIP, Western Blotting

TATA-box Binding Protein (TBP) is a general transcription factor that functions at the core of the DNA-binding multiprotein factor TFIID. Binding of TFIID to the TATA box is the initial transcriptional step of the pre-initiation complex (PIC). TBP plays a role in the activation of eukaryotic genes transcribed by RNA polymerase II and is a component of the transcription factor SL1/TIF-IB complex, which is involved in the assembly of the PIC (preinitiation complex) during RNA polymerase I-dependent transcription (Unitprot).
TRUSTED_SUSTAINABILITY

Specifications

Antigen TATA Binding Protein (TBP)α
Applications ChIP Assay, Western Blot
Classification Monoclonal
Conjugate Unconjugated
Formulation Anti-TATA Binding Protein (TBP) (mouse monoclonal). One vial containing 50μL ascites containing 0.05% sodium azide. Negative Ascites (mouse). One vial containing 50μL of mouse ascites with 0.05% sodium azide. ChIP Primers, GAPDH promoter. One vial containing 75μL of 5μM each primer specific for human GAPDH. FOR: TAC TAG CGG TTT TAC GGG CG; REV: TCG AAC AGG AGG AGC AGA GAG CGA
Gene Accession No. P20226
Gene Symbols TBP, MGC126055, SCA17, GTF2D1, TFIID, TF2D, MGC117320, GTF2D, MGC126054
Host Species Mouse
Immunogen Full length recombinant human TBP.
Purification Method Unpurified
Quantity 25 Assays
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_003185
Test Specificity Recognizes TATA Binding Protein (TBP).
Includes The ChIPAb+ TATA Binding Protein (TBP) set includes the TATA Binding Protein (TBP) antibody, a negative control mouse ascites and qPCR primers which amplify a 166bp region of human GAPDH promoter.
Target Species Chicken, Fish, Human, Mouse, Xenopus species
Content And Storage -20°C for one year from date of shipment; Upon first thaw and prior to removing the cap centrifuge the vial and gently mix the solution; Aliquot into microcentrifuge tubes and store at −20°C; Avoid repeated freeze/thaw cycles
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.