Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Upstate™ TCF-4α, Mouse monoclonal, ChIP Validated Antibody and Primer Set

Catalog No. 1710109MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 Assays
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
17-101-09MI 25 Assays Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-101-09MI Supplier MilliporeSigma™ Supplier No. 1710109
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Specifically detects TCF-4α Clone: in Human, Mouse samples, and it is validated for Chromatin Immunoprecipitation, Dot Blot, Western Blotting

TCF4, also known as TCF7L2, is expressed widely during development. Gene targeting study indicates that it is required to maintain the crypt stem cells of the small intestine. TCF4 has many different splicing isoforms and they are expressed differentially in tissues and in cancers of different stages. Studies also indicate that variant of the TCF4 gene confers an increased risk of type 2 diabetes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCF-4α
Applications ChIP Assay, Dot Blot, Western Blot
Classification Monoclonal
Conjugate Unconjugated
Formulation Anti-TCF-4 (mouse monoclonal). One vial containing 50μg of purified mouse monoclonal IgG2a in 50μL of buffer containing 0.1 M Tris-glycine, pH 7.4, 0.15 M NaCl, 0.05% sodium azide and 30% glycerol. Normal Mouse IgG. Two vials containing 25μg of purified IgG in 25μL of storage buffer containing 0.1% sodium azide. ChIP Primers, SP5 promote. One vial containing 75μL of 5μM of each primer specific for human SP5 promotor. FOR: GGG TCT CCA GGC GGC AAG; REV: AGC GAA AGC AAA TCC TTT GAA TCC
Gene Accession No. Q9NQB0
Gene Symbols TCF4, TCF-4, TCF7L2
Host Species Mouse
Immunogen Amino acids 31-331 of human TCF-4.
Purification Method Protein G chromatography
Quantity 25 Assays
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) CAB97213.1
Test Specificity This antibody recognizes TCF-4
Includes This ChIPAb+ TCF-4 -ChIP Validated Antibody and Primer Set conveniently includes the antibody and the specific control PCR primers.
Target Species Human, Mouse
Content And Storage -20°C for one year from date of shipment; Upon first thaw and prior to removing the cap centrifuge the vial and gently mix the solution; Aliquot into microcentrifuge tubes and store at −20°C; Avoid repeated freeze/thaw cycles
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.