Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Thrombin-Activatable Fibrinolysis Inhibitor, Human, Protein

Catalog No. 60519750UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-519-750UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-519-750UG Supplier MilliporeSigma™ Supplier No. 60519750UG
Only null left
Add to Cart
Add to Cart
Native thrombin-activatable fibrinolysis inhibitor from human liver. Single chain glycoprotein synthesized in the liver. Circulates in plasma as a zymogen at a concentration of 50nM. Activated by thrombin-thrombomodulin complex by proteolytic cleavage at Arg92. Believed to play an important role in interaction between the procoagulant, anticoagulant, and fibrinolytic systems.
TRUSTED_SUSTAINABILITY

Specifications

Concentration Concentration on Label
Formulation In 150mM NaCl, 20mM HEPES, pH 7.4.
Purification Method SDS-PAGE
Quantity 50 μg
Immunogen Thrombin-Activatable Fibrinolysis Inhibitor
Storage Requirements Following initial thaw, aliquot and freeze (−70°C).
Regulatory Status RUO
Species Human
Recombinant Native
Form Liquid
Purity or Quality Grade ≥95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.