Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Thrombin, Bovine, High Activity, Protein
Click to view available options
Quantity:
100 U
500 U
Description
Native, bovine thrombin. Key enzyme that activates factor XIII and converts fibronogen into fibrin.

Specifications
Specifications
Formulation | Lyophilized from 200mM NaCl, 50mM sodium citrate, 1mL 0.1% PEG, pH 6.5. |
Preparation Method | H2O |
Purification Method | Complete activation from homogeneous prothrombin by SDS-PAGE |
Quantity | 100 U |
Immunogen | Thrombin |
Storage Requirements | Following reconstitution, aliquot and freeze (−20°C). Stock solutions are stable for up to 2 months at −20°C. |
Regulatory Status | RUO |
Species | Bovine |
Recombinant | Native |
Form | Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction