Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Thrombopoietin Protein

Catalog No. GF037 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
GF037 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. GF037 Supplier MilliporeSigma™ Supplier No. GF037
Only null left
Add to Cart
Add to Cart

stimulates the proliferation and maturation of megakaryocytes and promotes increased circulating levels of platelets in vivo.

Megakaryocytopoiesis is the cellular development process that leads to platelet production. The protein encoded by this gene is a humoral growth factor that is necessary for megakaryocyte proliferation and maturation, as well as for thrombopoiesis. This protein is the ligand for MLP/C_MPL, the product of myeloproliferative leukemia virus oncogene.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Cell Culture
Quantity 10 μg
Form Lyophilized
Purity or Quality Grade Greater than 98% by SDS-PAGE/HPLC analyses
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.