Learn More
MilliporeSigma™ TLC Cellulose Aluminum Sheets
Catalog No. M1055520001 Shop All MilliporeSigma Products
Change view
Click to view available options
Dimensions (L x W):
20 x 20 cm
Catalog No. Dimensions (L x W)
M1055520001 20 x 20 cm
1 options
Catalog No. M1055520001 Supplier MilliporeSigma™ Supplier No. 1.05552.0001
Add to Cart
Add to Cart
25 aluminum sheets 20 x 20 cm
Our cellulose plates are used to analyze polar substances. An organic sorbent, cellulose is perfect for separating hydrophilic substances by partition chromatography. Typical applications include the analysis of amino acids, carbohydrates, and phosphates as well as nucleic acid and nucleic acid derivatives. We offer cellulose plates in two grades: TLC for conventional and HPTLC for demanding, high-performance separations.
TRUSTED_SUSTAINABILITY
Specifications
Backing Material Aluminum
Material Cellulose
Pore Size 60 Å
Quantity 25/Pk.
Type Cellulose TLC Plate
Fluorescent Indicator No
For Use With (Application) Analysis of Polar Substances, Separation of Hydrophilic Substances by Partition Chromatography, for Investigating in-Vitro Samples Derived From The Human Body Without Any Medical Objective
Sorbent Cellulose
Thickness (Metric) 70-110 μm
Dimensions (L x W) 20 x 20 cm
Length (Metric) 20 cm
Width (Metric) 20 cm